Product Details

SNP ID
rs149187785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:189003759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGATCCTTCTACTTTCCCAAAT[G/T]CTACCCTTTTAAAGACTCTTTGTGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TPRG1 PubMed Links

Gene Details

Gene
TPRG1
Gene Name
tumor protein p63 regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198485.3 Intron NP_940887.1
XM_005247382.4 Intron XP_005247439.1
XM_005247388.3 Intron XP_005247445.1
XM_011512732.2 Intron XP_011511034.2
XM_011512734.2 Intron XP_011511036.1
XM_011512735.2 Intron XP_011511037.1
XM_011512736.2 Intron XP_011511038.1
XM_017006257.1 Intron XP_016861746.1
XM_017006258.1 Intron XP_016861747.1
XM_017006259.1 Intron XP_016861748.1
XM_017006260.1 Intron XP_016861749.1
XM_017006261.1 Intron XP_016861750.1
XM_017006262.1 Intron XP_016861751.1
XM_017006263.1 Intron XP_016861752.1

View Full Product Details