Product Details

SNP ID
rs149353550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121595713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTAAAATCTCAAGAAGGCAAA[G/T]AGATTACTTTAATAATTATCTTTAT
Phenotype
MIM: 611164 MIM: 609107
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARGFX PubMed Links

Gene Details

Gene
ARGFX
Gene Name
arginine-fifty homeobox
There are no transcripts associated with this gene.

Gene
FBXO40
Gene Name
F-box protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016298.3 Intron NP_057382.2

View Full Product Details