Product Details

SNP ID
rs149415727
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160436431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAAAAGTCGTAACACATGCATC[A/G]TAACTCCAGTCCCTGTTAAAGGGAA
Phenotype
MIM: 605575 MIM: 616148
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SMC4 PubMed Links

Gene Details

Gene
SMC4
Gene Name
structural maintenance of chromosomes 4
There are no transcripts associated with this gene.

Gene
TRIM59
Gene Name
tripartite motif containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173084.2 2948 UTR 3 NP_775107.1

View Full Product Details