Product Details

SNP ID
rs149888444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185079821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCATATTACTGGGTCCAAAGTA[C/T]ACAGCTTTCATATCTGTCAGTCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf70 PubMed Links

Gene Details

Gene
C3orf70
Gene Name
chromosome 3 open reading frame 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025266.1 4130 UTR 3 NP_001020437.1

View Full Product Details