Product Details

SNP ID
rs150423919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49805941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTTTTCAGGTGACCATCCGGC[C/T]GCATAGAGCAGGGCTGAGCCGTGCA
Phenotype
MIM: 191325
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDHR4 PubMed Links

Gene Details

Gene
CDHR4
Gene Name
cadherin related family member 4
There are no transcripts associated with this gene.

Gene
FAM212A
Gene Name
family with sequence similarity 212 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203370.1 2864 Intron NP_976248.1
XM_017006372.1 2864 Intron XP_016861861.1
Gene
MIR5193
Gene Name
microRNA 5193
There are no transcripts associated with this gene.

Gene
UBA7
Gene Name
ubiquitin like modifier activating enzyme 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003335.2 2864 Silent Mutation GCA,GCG A955A NP_003326.2
XM_005265430.2 2864 Silent Mutation GCA,GCG A930A XP_005265487.1
XM_006713321.2 2864 Silent Mutation GCA,GCG A948A XP_006713384.1
XM_011534070.1 2864 Silent Mutation GCA,GCG A903A XP_011532372.1
XM_011534071.1 2864 Silent Mutation GCA,GCG A853A XP_011532373.1
XM_011534072.1 2864 Silent Mutation GCA,GCG A739A XP_011532374.1

View Full Product Details