Product Details

SNP ID
rs150659495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52080626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCATGTGAGCCATGAAAACTCA[A/G]TATTATAGGAACCGAAAGGACATAG
Phenotype
MIM: 614783
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POC1A PubMed Links

Gene Details

Gene
POC1A
Gene Name
POC1 centriolar protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161580.1 Intron NP_001155052.1
NM_001161581.1 Intron NP_001155053.1
NM_015426.4 Intron NP_056241.3
XM_011533560.1 Intron XP_011531862.1
XM_011533561.1 Intron XP_011531863.1
XM_011533562.2 Intron XP_011531864.1
XM_011533563.1 Intron XP_011531865.1
XM_011533564.2 Intron XP_011531866.1
XM_011533565.2 Intron XP_011531867.1
XM_017006104.1 Intron XP_016861593.1

View Full Product Details