Product Details

SNP ID
rs121912683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185145020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAACCTGGCGTCCGGTGGGGCCG[A/C]TGGGGCCACCTCCCTTTGCTTTGTC
Phenotype
MIM: 103220
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A4 PubMed Links

Gene Details

Gene
SLC25A4
Gene Name
solute carrier family 25 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001151.3 478 Missense Mutation GAT,GCT D123A NP_001142.2

View Full Product Details