Product Details

SNP ID
rs138037909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38971128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTGATCCAAAGGCAGCCTCTGC[A/G]TGGTCTCTGCTATTTAAGAAGGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM156 PubMed Links

Gene Details

Gene
TMEM156
Gene Name
transmembrane protein 156
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303228.1 986 Missense Mutation ACG,ATG T277M NP_001290157.1
NM_024943.2 986 Missense Mutation ACG,ATG T278M NP_079219.1
XM_005262670.3 986 Intron XP_005262727.1
XM_011513749.2 986 Missense Mutation ACG,ATG T278M XP_011512051.1
XM_011513750.2 986 Missense Mutation ACG,ATG T277M XP_011512052.1
XM_011513751.2 986 Intron XP_011512053.1
XM_011513752.2 986 Intron XP_011512054.1
XM_011513753.2 986 Missense Mutation ACG,ATG T250M XP_011512055.1
XM_011513754.2 986 Missense Mutation ACG,ATG T208M XP_011512056.1
XM_017008629.1 986 Missense Mutation ACG,ATG T208M XP_016864118.1

View Full Product Details