Product Details

SNP ID
rs138163101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:107614250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGGAGTGACTGGGTTAACAGCC[A/T]AAGCTTTCTCCAAGGATTTGTAGTA
Phenotype
MIM: 603262
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PAPSS1 PubMed Links

Gene Details

Gene
PAPSS1
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 1
There are no transcripts associated with this gene.

View Full Product Details