Product Details

SNP ID
rs138619142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122893015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGAGTTGTATTTTCAGTCTTC[A/G]CCAGGTCATTGAGATCCATCCACTC
Phenotype
MIM: 134920 MIM: 606261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF2 PubMed Links

Gene Details

Gene
FGF2
Gene Name
fibroblast growth factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002006.4 730 UTR 3 NP_001997.5
Gene
NUDT6
Gene Name
nudix hydrolase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007083.4 730 Missense Mutation GCG,GTG A255V NP_009014.2
NM_198041.2 730 Missense Mutation GCG,GTG A86V NP_932158.1

View Full Product Details