Product Details

SNP ID
rs138941384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17799798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTTTCTTAGGAAGCAGATGG[G/T]ATTAAAAGACGTGATTATGGTCTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017741.3 5289 Intron NP_060211.3
XM_005248169.3 5289 UTR 3 XP_005248226.1
XM_005248171.3 5289 UTR 3 XP_005248228.1
XM_006713966.3 5289 UTR 3 XP_006714029.1
XM_017008327.1 5289 UTR 3 XP_016863816.1
XM_017008328.1 5289 UTR 3 XP_016863817.1

View Full Product Details