Product Details

SNP ID
rs139089250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68477580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGATGCATCCTATGAGTTTCAA[C/G]CAGGTGATGTGATGTTTGTGACAGG
Phenotype
MIM: 610399
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11E PubMed Links

Gene Details

Gene
TMPRSS11E
Gene Name
transmembrane protease, serine 11E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014058.3 983 Missense Mutation CCA,GCA P307A NP_054777.2
XM_011531896.2 983 Missense Mutation CCA,GCA P229A XP_011530198.1

View Full Product Details