Product Details

SNP ID
rs139368028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102509760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCTTTCTTCTTCAGTCAGTCA[C/T]CAAGTTCTATCAAGGCAGCCTCCTC
Phenotype
MIM: 164011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105377621 PubMed Links

Gene Details

Gene
LOC105377621
Gene Name
uncharacterized LOC105377621
There are no transcripts associated with this gene.

Gene
NFKB1
Gene Name
nuclear factor kappa B subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165412.1 Intron NP_001158884.1
NM_001319226.1 Intron NP_001306155.1
NM_003998.3 Intron NP_003989.2
XM_011532006.2 Intron XP_011530308.1
XM_011532008.2 Intron XP_011530310.1

View Full Product Details