Product Details

SNP ID
rs139575168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70688992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCGGGGGAGGAGGAGGAGGAGGA[G/T]AATGCCGATGATGATGGTGGGAGCT
Phenotype
MIM: 611614
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UTP3 PubMed Links

Gene Details

Gene
UTP3
Gene Name
UTP3, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020368.2 514 Silent Mutation GAG,GAT E105D NP_065101.1

View Full Product Details