Product Details

SNP ID
rs139866800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:988870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGGTGGCCTCCAGCTCCCTGTGG[C/T]GGGCTCGTGCTGTCTGCACGGCATC
Phenotype
MIM: 252800 MIM: 610130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IDUA PubMed Links

Gene Details

Gene
IDUA
Gene Name
iduronidase, alpha-L-
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000203.4 2236 Intron NP_000194.2
XM_011513461.2 2236 Intron XP_011511763.1
XM_017008163.1 2236 Intron XP_016863652.1
Gene
SLC26A1
Gene Name
solute carrier family 26 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022042.3 2236 Missense Mutation CAC,CGC H690R NP_071325.2
NM_134425.2 2236 Intron NP_602297.1
NM_213613.3 2236 Missense Mutation CAC,CGC H690R NP_998778.1

View Full Product Details