Product Details
- SNP ID
-
rs140087355
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:500536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCTACACAACTTACCTTGTGGAA[A/G]AAGGAGCATCTCACAGTTTTGTGGC
- Phenotype
-
MIM: 616918
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIGG
PubMed Links
Gene Details
- Gene
- PIGG
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class G
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001127178.2 |
665 |
Missense Mutation |
AAA,GAA |
K99E |
NP_001120650.1 |
NM_001289051.1 |
665 |
Missense Mutation |
AAA,GAA |
K10E |
NP_001275980.1 |
NM_001289052.1 |
665 |
Missense Mutation |
AAA,GAA |
K99E |
NP_001275981.1 |
NM_001289053.1 |
665 |
Missense Mutation |
AAA,GAA |
K10E |
NP_001275982.1 |
NM_001289055.1 |
665 |
UTR 5 |
|
|
NP_001275984.1 |
NM_001289057.1 |
665 |
Missense Mutation |
AAA,GAA |
K10E |
NP_001275986.1 |
NM_017733.4 |
665 |
Missense Mutation |
AAA,GAA |
K99E |
NP_060203.3 |
XM_005272284.2 |
665 |
Missense Mutation |
AAA,GAA |
K10E |
XP_005272341.1 |
XM_011513490.2 |
665 |
Missense Mutation |
AAA,GAA |
K99E |
XP_011511792.1 |
XM_011513491.1 |
665 |
UTR 5 |
|
|
XP_011511793.1 |
XM_011513492.1 |
665 |
Intron |
|
|
XP_011511794.1 |
XM_011513493.1 |
665 |
Missense Mutation |
AAA,GAA |
K99E |
XP_011511795.1 |
XM_011513494.2 |
665 |
Missense Mutation |
AAA,GAA |
K99E |
XP_011511796.1 |
XM_017008321.1 |
665 |
Missense Mutation |
AAA,GAA |
K10E |
XP_016863810.1 |
XM_017008322.1 |
665 |
UTR 5 |
|
|
XP_016863811.1 |
XM_017008323.1 |
665 |
UTR 5 |
|
|
XP_016863812.1 |
XM_017008324.1 |
665 |
UTR 5 |
|
|
XP_016863813.1 |
XM_017008325.1 |
665 |
UTR 5 |
|
|
XP_016863814.1 |
XM_017008326.1 |
665 |
Missense Mutation |
AAA,GAA |
K99E |
XP_016863815.1 |
- Gene
- ZNF721
- Gene Name
- zinc finger protein 721
There are no transcripts associated with this gene.
View Full Product Details