Product Details

SNP ID
rs140222606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186534171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGAGGAAGTGGAAAACCACCA[C/T]GGCGATGGTGTAGGCGGAGCTGACG
Phenotype
MIM: 600665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTNR1A PubMed Links

Gene Details

Gene
MTNR1A
Gene Name
melatonin receptor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005958.4 773 Missense Mutation ATG,GTG M191V NP_005949.1
XM_011532002.2 773 Missense Mutation ATG,GTG M106V XP_011530304.1

View Full Product Details