Product Details

SNP ID
rs140708408
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128271319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTTGGTTGTTTACAAAGTTCA[A/G]TCCTGTTTATTGTGATCCTTGGTAT
Phenotype
MIM: 607735
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PGRMC2 PubMed Links

Gene Details

Gene
PGRMC2
Gene Name
progesterone receptor membrane component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006320.4 1708 Silent Mutation GAC,GAT D247D NP_006311.2
XM_011531533.1 1708 Intron XP_011529835.1

View Full Product Details