Product Details
- SNP ID
-
rs140790882
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:54100512 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGCTGCCCGTCCCGCAAGAGCGG[C/T]GCGTTCTGCGTGTGCCCTCTCTGCG
- Phenotype
-
MIM: 616253
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSX2
PubMed Links
Gene Details
- Gene
- GSX2
- Gene Name
- GS homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_133267.2 |
432 |
Silent Mutation |
GGC,GGT |
G56G |
NP_573574.1 |
View Full Product Details