Product Details

SNP ID
rs141053214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183664018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCGGAGAGCTGATCGAGTACCA[A/G]TTTCTTTCAAGGTGCTCCCAGGTGA
Phenotype
MIM: 614138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RWDD4 PubMed Links

Gene Details

Gene
RWDD4
Gene Name
RWD domain containing 4
There are no transcripts associated with this gene.

Gene
TRAPPC11
Gene Name
trafficking protein particle complex 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021942.5 353 Missense Mutation ATT,GTT I51V NP_068761.4
NM_199053.2 353 Missense Mutation ATT,GTT I51V NP_951008.1
XM_017008537.1 353 Missense Mutation ATT,GTT I51V XP_016864026.1
XM_017008538.1 353 Missense Mutation ATT,GTT I51V XP_016864027.1
XM_017008539.1 353 UTR 5 XP_016864028.1

View Full Product Details