Product Details

SNP ID
rs141328282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73572323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAAGCCACAGTGACTTTTATGA[C/T]CTACTTCAAAGTCTCACAGTCTCTT
Phenotype
MIM: 612620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASSF6 PubMed Links

Gene Details

Gene
RASSF6
Gene Name
Ras association domain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270391.1 7779 UTR 3 NP_001257320.1
NM_001270392.1 7779 UTR 3 NP_001257321.1
NM_177532.4 7779 UTR 3 NP_803876.1
NM_201431.2 7779 UTR 3 NP_958834.1
XM_011531687.2 7779 Intron XP_011529989.1
XM_011531688.1 7779 UTR 3 XP_011529990.1
XM_011531689.2 7779 UTR 3 XP_011529991.1
XM_011531690.2 7779 UTR 3 XP_011529992.1
XM_011531691.1 7779 UTR 3 XP_011529993.1
XM_017007837.1 7779 UTR 3 XP_016863326.1
XM_017007838.1 7779 Intron XP_016863327.1

View Full Product Details