Product Details

SNP ID
rs141459493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165079659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCTGTCATGACCGTGAGCCTCT[C/G]ACGTTCTACTTGGCTGACAGCCCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM192 PubMed Links

Gene Details

Gene
TMEM192
Gene Name
transmembrane protein 192
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011531717.2 966 UTR 3 XP_011530019.1

View Full Product Details