Product Details

SNP ID
rs141657790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144646880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGAGTGGGGGAGAGATGCTG[C/T]GCGGTGGCTTCTACCCTCGGCTGTC
Phenotype
MIM: 606178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HHIP PubMed Links

Gene Details

Gene
HHIP
Gene Name
hedgehog interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022475.2 754 Missense Mutation CGC,TGC R69C NP_071920.1
XM_005263178.4 754 Missense Mutation CGC,TGC R69C XP_005263235.1
XM_006714288.3 754 Missense Mutation CGC,TGC R69C XP_006714351.1
Gene
HHIP-AS1
Gene Name
HHIP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details