Product Details

SNP ID
rs142071292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48488477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGTGGTCCTTTTCATAATGAC[C/T]GGCACTATGTTAGGACCTGAACTGC
Phenotype
MIM: 607520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRYL PubMed Links

Gene Details

Gene
FRYL
Gene Name
FRY like transcription coactivator
There are no transcripts associated with this gene.

Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152679.3 1071 Silent Mutation ACC,ACT T284T NP_689892.1
Gene
ZAR1
Gene Name
zygote arrest 1
There are no transcripts associated with this gene.

View Full Product Details