Product Details
- SNP ID
-
rs142258660
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:55080024 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACCGGTTTGCACTCCAATCTCTA[A/T]CAGCTTTAAAAGTTCTGCTTCCTCA
- Phenotype
-
MIM: 191306
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KDR
PubMed Links
Gene Details
- Gene
- KDR
- Gene Name
- kinase insert domain receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002253.2 |
4290 |
Missense Mutation |
ATA,TTA |
I1330L |
NP_002244.1 |
View Full Product Details