Product Details

SNP ID
rs142735873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76436120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAACTCTAACACGGCCACCAAGG[A/G]AAGGTACATTTATCTGGAGGCATTC
Phenotype
MIM: 604570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHROOM3 PubMed Links

Gene Details

Gene
SHROOM3
Gene Name
shroom family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020859.3 819 Missense Mutation GAA,GGA E23G NP_065910.3
XM_005263163.4 819 Intron XP_005263220.1
XM_005263164.4 819 Intron XP_005263221.1
XM_005263165.4 819 Intron XP_005263222.1
XM_006714282.3 819 Intron XP_006714345.1
XM_011532157.2 819 Intron XP_011530459.2
XM_011532158.2 819 Missense Mutation GAA,GGA E23G XP_011530460.1
XM_011532159.1 819 Missense Mutation GAA,GGA E23G XP_011530461.1
XM_011532160.1 819 Intron XP_011530462.1
XM_011532161.2 819 Intron XP_011530463.1
XM_017008488.1 819 Intron XP_016863977.1
XM_017008489.1 819 Intron XP_016863978.1

View Full Product Details