Product Details
- SNP ID
-
rs142984801
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:69999751 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGTCCTCCCTACATTTGTATTGA[A/T]TTATTAAACTTTTCTTCATTTTTCA
- Phenotype
-
MIM: 184470
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
STATH
PubMed Links
Gene Details
- Gene
- STATH
- Gene Name
- statherin
View Full Product Details