Product Details

SNP ID
rs143745435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:108625164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCTTATCGAGGAGCAGAAAAT[C/T]GTTGTGAAAGTGTTGAAGGCACAAG
Phenotype
MIM: 616862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL34 PubMed Links

Gene Details

Gene
RPL34
Gene Name
ribosomal protein L34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000995.4 358 Silent Mutation ATC,ATT I102I NP_000986.2
NM_001319232.1 358 Intron NP_001306161.1
NM_001319234.1 358 Silent Mutation ATC,ATT I102I NP_001306163.1
NM_001319235.1 358 Intron NP_001306164.1
NM_001319236.1 358 Intron NP_001306165.1
NM_033625.3 358 Silent Mutation ATC,ATT I102I NP_296374.1
Gene
RPL34-AS1
Gene Name
RPL34 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details