Product Details

SNP ID
rs143977959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:79842649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGAAATGTTATTCTTGTAGCA[A/C]CATTGTCTTTCTTCTTTTATAATAC
Phenotype
MIM: 609717
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCAT4 PubMed Links

Gene Details

Gene
PCAT4
Gene Name
prostate cancer associated transcript 4 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details