Product Details
- SNP ID
-
rs144339935
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:56966659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCTGCATGCTTCTGATACAGAGC[A/G]TCTGCCCTGTCCAAGGAGGTGATAT
- Phenotype
-
MIM: 614919
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOA1
PubMed Links
Gene Details
- Gene
- NOA1
- Gene Name
- nitric oxide associated 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032313.3 |
1800 |
Silent Mutation |
GAC,GAT |
D575D |
NP_115689.1 |
View Full Product Details