Product Details

SNP ID
rs144389166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68477528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCTTTCTAGCCCTGTTCCCTA[C/T]ACAAATGCAGTACATAGAGTTTGTC
Phenotype
MIM: 610399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11E PubMed Links

Gene Details

Gene
TMPRSS11E
Gene Name
transmembrane protease, serine 11E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014058.3 931 Silent Mutation TAC,TAT Y289Y NP_054777.2
XM_011531896.2 931 Silent Mutation TAC,TAT Y211Y XP_011530198.1

View Full Product Details