Product Details

SNP ID
rs144390629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73854066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAAGAATGGGAGGAAAATTTGC[A/T]TGGATCTGCAAGCCCTGCTGTACAA
Phenotype
MIM: 173461
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PF4V1 PubMed Links

Gene Details

Gene
PF4V1
Gene Name
platelet factor 4 variant 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002620.3 326 Missense Mutation ATG,TTG M87L NP_002611.1

View Full Product Details