Product Details

SNP ID
rs144471150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68496674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGAGGACCACTGGTTAGTTCAG[A/T]TGCTAGAGATATCTGGTACCTTGCT
Phenotype
MIM: 610399
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11E PubMed Links

Gene Details

Gene
TMPRSS11E
Gene Name
transmembrane protease, serine 11E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014058.3 1206 Missense Mutation GAT,GTT D381V NP_054777.2
XM_011531896.2 1206 Missense Mutation GAT,GTT D303V XP_011530198.1

View Full Product Details