Product Details
- SNP ID
-
rs145423393
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:79406987 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGGTTCATGGCTTCCAAAATCTCT[C/T]GGGTTTGGAAACAAACAGCTTCTAA
- Phenotype
-
MIM: 600148
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GK2
PubMed Links
Gene Details
- Gene
- GK2
- Gene Name
- glycerol kinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033214.2 |
1232 |
Missense Mutation |
|
|
NP_149991.2 |
View Full Product Details