Product Details

SNP ID
rs145842147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183663984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCATCGAGCTGTCTGGGACGC[C/G]TTCTGTGCAAATCGGAGAGCTGATC
Phenotype
MIM: 614138
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RWDD4 PubMed Links

Gene Details

Gene
RWDD4
Gene Name
RWD domain containing 4
There are no transcripts associated with this gene.

Gene
TRAPPC11
Gene Name
trafficking protein particle complex 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021942.5 319 Silent Mutation GCC,GCG A39A NP_068761.4
NM_199053.2 319 Silent Mutation GCC,GCG A39A NP_951008.1
XM_017008537.1 319 Silent Mutation GCC,GCG A39A XP_016864026.1
XM_017008538.1 319 Silent Mutation GCC,GCG A39A XP_016864027.1
XM_017008539.1 319 UTR 5 XP_016864028.1

View Full Product Details