Product Details

SNP ID
rs145910626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128272510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCTACCAGCAAATATTCCATA[C/T]GGACCCGCTGGAAAAAAGAAAATAA
Phenotype
MIM: 607735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGRMC2 PubMed Links

Gene Details

Gene
PGRMC2
Gene Name
progesterone receptor membrane component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006320.4 1668 Silent Mutation CCA,CCG P166P NP_006311.2
XM_011531533.1 1668 UTR 3 XP_011529835.1

View Full Product Details