Product Details

SNP ID
rs146247849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73576292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTGAAGACATTTCAGTATAAT[C/T]GCCTTTTCTTTATTGAATCTGAAAA
Phenotype
MIM: 612620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASSF6 PubMed Links

Gene Details

Gene
RASSF6
Gene Name
Ras association domain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270391.1 3810 Silent Mutation GCA,GCG A285A NP_001257320.1
NM_001270392.1 3810 Silent Mutation GCA,GCG A307A NP_001257321.1
NM_177532.4 3810 Silent Mutation GCA,GCG A319A NP_803876.1
NM_201431.2 3810 Silent Mutation GCA,GCG A351A NP_958834.1
XM_011531687.2 3810 UTR 3 XP_011529989.1
XM_011531688.1 3810 Silent Mutation GCA,GCG A319A XP_011529990.1
XM_011531689.2 3810 Silent Mutation GCA,GCG A317A XP_011529991.1
XM_011531690.2 3810 Silent Mutation GCA,GCG A261A XP_011529992.1
XM_011531691.1 3810 Silent Mutation GCA,GCG A261A XP_011529993.1
XM_017007837.1 3810 Silent Mutation GCA,GCG A223A XP_016863326.1
XM_017007838.1 3810 Intron XP_016863327.1

View Full Product Details