Product Details

SNP ID
rs146591833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:137521221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCACACTGGAGTGAGTTCCAAG[C/T]GGCGGCCCACAGTTGGTGGTGGGAT
Phenotype
MIM: 608287
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH18 PubMed Links

Gene Details

Gene
PCDH18
Gene Name
protocadherin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300828.1 3626 Silent Mutation CCA,CCG P1071P NP_001287757.1
NM_019035.4 3626 Silent Mutation CCA,CCG P1072P NP_061908.1
XM_006714239.3 3626 Intron XP_006714302.1
XM_017008311.1 3626 UTR 3 XP_016863800.1

View Full Product Details