Product Details

SNP ID
rs147011953
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:163525571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTTCTTTCTTTTACAATTTAAT[A/C]TTTACTCCTGTACGATAAGAGTGAT
Phenotype
MIM: 613331
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MARCH1 PubMed Links

Gene Details

Gene
MARCH1
Gene Name
membrane associated ring-CH-type finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166373.1 4116 UTR 3 NP_001159845.1
NM_017923.3 4116 UTR 3 NP_060393.1
XM_011532055.2 4116 UTR 3 XP_011530357.1
XM_017008334.1 4116 UTR 3 XP_016863823.1
XM_017008335.1 4116 UTR 3 XP_016863824.1
XM_017008336.1 4116 UTR 3 XP_016863825.1
Gene
TMA16
Gene Name
translation machinery associated 16 homolog
There are no transcripts associated with this gene.

View Full Product Details