Product Details

SNP ID
rs147669408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:988932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACAGCTGCTCCTCCTCAGCCGT[C/G]TCCCCGGGGCCCTCCCCGAGGAAGC
Phenotype
MIM: 252800 MIM: 610130
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IDUA PubMed Links

Gene Details

Gene
IDUA
Gene Name
iduronidase, alpha-L-
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000203.4 2174 Intron NP_000194.2
XM_011513461.2 2174 Intron XP_011511763.1
XM_017008163.1 2174 Intron XP_016863652.1
Gene
SLC26A1
Gene Name
solute carrier family 26 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022042.3 2174 Missense Mutation GAC,GAG D669E NP_071325.2
NM_134425.2 2174 Intron NP_602297.1
NM_213613.3 2174 Missense Mutation GAC,GAG D669E NP_998778.1

View Full Product Details