Product Details

SNP ID
rs147745691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17803553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGTCAGTGTAAGAATAAGTAG[C/T]GTTGATGGGTTCAGTACGAGTTGTT
Phenotype
MIM: 606280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017741.3 1534 Missense Mutation ACT,GCT T197A NP_060211.3
XM_005248169.3 1534 Missense Mutation ACT,GCT T197A XP_005248226.1
XM_005248171.3 1534 Missense Mutation ACT,GCT T197A XP_005248228.1
XM_006713966.3 1534 Missense Mutation ACT,GCT T197A XP_006714029.1
XM_017008327.1 1534 Missense Mutation ACT,GCT T197A XP_016863816.1
XM_017008328.1 1534 Missense Mutation ACT,GCT T197A XP_016863817.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
There are no transcripts associated with this gene.

View Full Product Details