Product Details

SNP ID
rs147893499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:150083741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTAAAGTCATGCATGGTGATTG[A/G]ACTTGGAAAGTTAATATTGAATTTC
Phenotype
MIM: 613166
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCLK2 PubMed Links

Gene Details

Gene
DCLK2
Gene Name
doublecortin like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040260.3 Intron NP_001035350.2
NM_001040261.4 Intron NP_001035351.4
XM_005262785.2 Intron XP_005262842.1
XM_005262786.2 Intron XP_005262843.1
XM_017007832.1 Intron XP_016863321.1
XM_017007833.1 Intron XP_016863322.1

View Full Product Details