Product Details

SNP ID
rs149101873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185145219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAGGCATCATTATCTATAGAGC[C/T]GCCTACTTCGGAGTCTATGATACTG
Phenotype
MIM: 103220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A4 PubMed Links

Gene Details

Gene
SLC25A4
Gene Name
solute carrier family 25 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001151.3 677 Silent Mutation GCC,GCT A189A NP_001142.2

View Full Product Details