Product Details

SNP ID
rs149186367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82753367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGCACTGCTCCTGTCCAACTG[A/G]CTCATTGTGCAGACAGCCTGAGTAG
Phenotype
MIM: 608370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR575 PubMed Links

Gene Details

Gene
MIR575
Gene Name
microRNA 575
There are no transcripts associated with this gene.

Gene
SCD5
Gene Name
stearoyl-CoA desaturase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037582.2 Intron NP_001032671.2
NM_024906.2 Intron NP_079182.2

View Full Product Details