Product Details

SNP ID
rs149200314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:849920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGGGGTGCACAGCCAGCACCGCG[C/T]GGCGATAGTGCTTCTTCACTTGCTC
Phenotype
MIM: 602052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAK PubMed Links

Gene Details

Gene
GAK
Gene Name
cyclin G associated kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318134.1 4081 Missense Mutation NP_001305063.1
NM_005255.3 4081 Missense Mutation NP_005246.2
XM_005272268.2 4081 Missense Mutation XP_005272325.1
XM_005272270.2 4081 Missense Mutation XP_005272327.1
XM_011513425.2 4081 Missense Mutation XP_011511727.1
XM_011513426.2 4081 Missense Mutation XP_011511728.1
XM_011513427.2 4081 Missense Mutation XP_011511729.1
XM_011513428.2 4081 Missense Mutation XP_011511730.1
XM_011513429.2 4081 Missense Mutation XP_011511731.1
XM_011513430.1 4081 Missense Mutation XP_011511732.1
XM_011513431.2 4081 Missense Mutation XP_011511733.1
XM_011513432.2 4081 Missense Mutation XP_011511734.1
XM_011513434.2 4081 Missense Mutation XP_011511736.1
XM_017007991.1 4081 Missense Mutation XP_016863480.1
XM_017007992.1 4081 Missense Mutation XP_016863481.1
XM_017007993.1 4081 Missense Mutation XP_016863482.1
XM_017007994.1 4081 Intron XP_016863483.1
XM_017007995.1 4081 Missense Mutation XP_016863484.1

View Full Product Details