Product Details
- SNP ID
-
rs149332468
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:83416763 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGAAACAATTTGCTTGGCTTGGC[A/G]TCTTGATAAATGATCAATTGTCCTT
- Phenotype
-
MIM: 606769
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HELQ
PubMed Links
Gene Details
- Gene
- HELQ
- Gene Name
- helicase, POLQ-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001297755.1 |
3016 |
Missense Mutation |
CGC,TGC |
R989C |
NP_001284684.1 |
NM_001297756.1 |
3016 |
Missense Mutation |
CGC,TGC |
R559C |
NP_001284685.1 |
NM_001297757.1 |
3016 |
Missense Mutation |
CGC,TGC |
R512C |
NP_001284686.1 |
NM_001297758.1 |
3016 |
Intron |
|
|
NP_001284687.1 |
NM_001297759.1 |
3016 |
Intron |
|
|
NP_001284688.1 |
NM_133636.3 |
3016 |
Missense Mutation |
CGC,TGC |
R1056C |
NP_598375.2 |
XM_005262711.1 |
3016 |
Missense Mutation |
CGC,TGC |
R1019C |
XP_005262768.1 |
XM_005262713.2 |
3016 |
Missense Mutation |
CGC,TGC |
R950C |
XP_005262770.1 |
XM_006714076.2 |
3016 |
Intron |
|
|
XP_006714139.1 |
XM_011531580.2 |
3016 |
Intron |
|
|
XP_011529882.1 |
XM_017007679.1 |
3016 |
Missense Mutation |
CGC,TGC |
R952C |
XP_016863168.1 |
XM_017007680.1 |
3016 |
Missense Mutation |
CGC,TGC |
R883C |
XP_016863169.1 |
XM_017007681.1 |
3016 |
Intron |
|
|
XP_016863170.1 |
XM_017007682.1 |
3016 |
Missense Mutation |
CGC,TGC |
R559C |
XP_016863171.1 |
XM_017007683.1 |
3016 |
Missense Mutation |
CGC,TGC |
R559C |
XP_016863172.1 |
View Full Product Details