Product Details

SNP ID
rs149433021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38827136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGTGTTAATTTCATATTAAAAG[C/T]GGCTCTAATGTTAGCCCAAAAGAGC
Phenotype
MIM: 605403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLR6 PubMed Links

Gene Details

Gene
TLR6
Gene Name
toll like receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006068.4 2444 Missense Mutation ACT,GCT T780A NP_006059.2
XM_005262637.4 2444 Missense Mutation ACT,GCT T780A XP_005262694.1
XM_011513612.2 2444 Missense Mutation ACT,GCT T780A XP_011511914.1
XM_011513613.2 2444 Missense Mutation ACT,GCT T780A XP_011511915.1
XM_011513614.2 2444 Missense Mutation ACT,GCT T780A XP_011511916.1

View Full Product Details