Product Details

SNP ID
rs149499063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184629486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCAGGAGCCATCCTTTGAATTT[C/T]GCCAAGAATAATAACCTAAAAGGCA
Phenotype
MIM: 600636
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP3 PubMed Links

Gene Details

Gene
CASP3
Gene Name
caspase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004346.3 883 Missense Mutation CAA,CGA Q207R NP_004337.2
NM_032991.2 883 Missense Mutation CAA,CGA Q207R NP_116786.1
XM_011532301.1 883 Missense Mutation CAA,CGA Q207R XP_011530603.1
Gene
LVCAT8
Gene Name
liver cancer-associated transcript 8
There are no transcripts associated with this gene.

View Full Product Details