Product Details

SNP ID
rs149620875
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186533911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACGCCATGTAGTAACTGGCCAC[A/C]AACAGCCACTCTGGGATCCTAGGCA
Phenotype
MIM: 600665
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MTNR1A PubMed Links

Gene Details

Gene
MTNR1A
Gene Name
melatonin receptor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005958.4 1033 Missense Mutation TTG,TTT L277F NP_005949.1
XM_011532002.2 1033 Missense Mutation TTG,TTT L192F XP_011530304.1

View Full Product Details