Product Details

SNP ID
rs149891396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68478870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGTTACAGTCAAAATCATCTTC[A/G]ACAAGCACAGGTGACTCTCATAGAC
Phenotype
MIM: 610399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11E PubMed Links

Gene Details

Gene
TMPRSS11E
Gene Name
transmembrane protease, serine 11E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014058.3 1053 Missense Mutation CAA,CGA Q330R NP_054777.2
XM_011531896.2 1053 Missense Mutation CAA,CGA Q252R XP_011530198.1

View Full Product Details